Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 3 | 569 | ||
NM_001271532.1 | NP_001258461.1 | 2 | 436 | |||
NM_133748.2 | NP_598509.1 | 3 | 812 | |||
NM_178082.3 | NP_835183.1 | 3 | 576 | |||
GenBank mRNA | AF527631.1 | - | 3 | 546 | ||
AK040703.1 | - | 3 | 567 | |||
AK050394.1 | - | 3 | 570 | |||
AK152076.1 | - | 3 | 542 | |||
AK154543.1 | - | 3 | 575 | |||
AK161256.1 | - | 3 | 580 | |||
AK165634.1 | - | 3 | 581 | |||
AK190310.1 | - | 1 | 47 | |||
BC015288.1 | - | 3 | 526 | 4240711 | ||
BC023067.1 | - | 3 | 550 | 5357599 | ||
BC023874.1 | - | 2 | 468 | 5355039 | ||
BC024411.1 | - | 3 | 564 | 4971297 |