Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028431.2 | NP_082707.1 | 12 | 1391 | ||
GenBank mRNA | AK013995.1 | - | 12 | 1392 | ||
AK152673.1 | - | 12 | 1393 | |||
AK169417.1 | - | 12 | 1348 | |||
BC038321.1 | - | 11 | 1262 | |||
BC051216.1 | - | 11 | 1198 | |||
BC060662.1 | - | 11 | 3903 | |||
BC065095.1 | - | 11 | 3903 | |||
BC087943.1 | - | 12 | 1403 |