Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285852.1 | NP_001272781.1 | 10, 11 | 995 | ||
NM_001285859.1 | NP_001272788.1 | 9, 10 | 908 | |||
NM_001285860.1 | NP_001272789.1 | 9, 10 | 908 | |||
NM_144906.2 | NP_659155.1 | 9, 10 | 911 | |||
GenBank mRNA | AB262964.1 | - | 9, 10 | 574 | ||
AK032439.1 | - | 9, 10 | 699 | |||
AK043018.1 | - | 9, 10 | 734 | |||
BC017596.1 | - | 9, 10 | 683 | 4482298 |