Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291069.1 | NP_001277998.1 | 10 | 1819 | ||
NM_175193.6 | NP_780402.1 | 9 | 1735 | |||
GenBank mRNA | AK033730.1 | - | 9 | 1722 | ||
AK048434.1 | - | 10 | 1646 | |||
AK078928.1 | - | 10 | 1819 | |||
BC086642.1 | - | 9 | 1567 | 6836803 | ||
BC089354.1 | - | 9 | 1617 | 6837211 |