Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198100.2 | NP_932768.2 | 2 | 365 | ||
GenBank mRNA | AK014173.1 | - | 3 | 327 | ||
BC057590.1 | - | 2 | 365 | 6810986 |