Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175195.3 | NP_780404.2 | 6 | 674 | ||
GenBank mRNA | AK046804.1 | - | 6 | 693 | ||
AK148109.1 | - | 6 | 641 | |||
AK157906.1 | - | 6 | 619 | |||
AK165324.1 | - | 6 | 799 | |||
AK166364.1 | - | 6 | 625 | |||
AK170375.1 | - | 6 | 640 | |||
AK197828.1 | - | 4 | 347 | |||
BC034206.1 | - | 3 | 229 | |||
BC052094.1 | - | 4 | 348 |