Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddit4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030143.4 | NP_084419.2 | 3 | 789 | ||
GenBank mRNA | AF327512.1 | - | 2 | 544 | ||
AF335325.1 | - | 2 | 589 | |||
AK029362.1 | - | 1 | 357 | |||
AK053333.1 | - | 3 | 727 | |||
BC038131.1 | - | 3 | 688 | |||
BC147181.1 | - | 2 | 568 | 9056209 | ||
BC147182.1 | - | 2 | 568 | 9056220 |