Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290783.1 | NP_001277712.1 | 2 | 765 | ||
NM_026137.5 | NP_080413.2 | 2 | 799 | |||
NM_028542.1 | NP_082818.1 | 2 | 1051 | |||
GenBank mRNA | AF220146.1 | - | 1 | 321 | ||
AF353243.1 | - | 2 | 445 | |||
AF513713.1 | - | 2 | 268 | |||
AK006837.1 | - | 1 | 287 | |||
AK017549.1 | - | 2 | 381 | |||
BC037502.1 | - | 2 | 416 | |||
BC057366.1 | - | 2 | 765 | |||
BC080853.1 | - | 2 | 714 | |||
BC108332.1 | - | 2 | 555 | 6493226 | ||
BC120553.1 | - | 2 | 302 | 40129476 | ||
BC125287.1 | - | 2 | 301 | 40129802 |