Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iws1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173441.3 | NP_775617.1 | 11 | 2233 | ||
GenBank mRNA | AK006925.1 | - | 5 | 526 | ||
AK028247.1 | - | 11 | 2204 | |||
AK033013.1 | - | 11 | 2228 | |||
AK136492.1 | - | 7 | 689 | |||
BC068184.1 | - | 10 | 2819 | 30639737 |