Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028597.3 | NP_082873.2 | 2 | 422 | ||
GenBank mRNA | AK010668.1 | - | 2 | 402 | ||
AK182377.1 | - | 1 | 39 | |||
AK214286.1 | - | 1 | 39 | |||
BC019603.1 | - | 2 | 396 | 2647864 |