Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021019.4 | NP_066299.2 | 4 | 404 | ||
NM_079423.3 | NP_524147.2 | 4 | 404 | |||
GenBank mRNA | AB046613.1 | - | 4 | 303 | ||
AK301198.1 | - | 3 | 582 | |||
BC006781.2 | - | 2 | 234 | |||
BC017455.1 | - | 4 | 291 | 3538373 | ||
BC071661.1 | - | 4 | 287 | 5561575 | ||
BC093066.1 | - | 4 | 300 | 30722094 | ||
BG283793.1 | - | 4 | 403 | |||
CN401857.1 | - | 4 | 288 | |||
CR456839.1 | - | 2 | 158 | |||
CR457014.1 | - | 3 | 263 | |||
M22918.1 | - | 4 | 294 | |||
M22920.1 | - | 3 | 272 | |||
M31212.1 | - | 4 | 303 | |||
U02629.1 | - | 4 | 301 |