Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080559.1 | NP_542126.1 | 2 | 283 | ||
GenBank mRNA | AK003431.1 | - | 2 | 264 | ||
AK179341.1 | - | 2 | 168 | |||
BC008110.1 | - | 2 | 283 | 3481987 |