Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290662.1 | NP_001277591.1 | 3 | 295 | ||
NM_134471.4 | NP_608301.3 | 3 | 336 | |||
GenBank mRNA | AK015046.1 | - | 3 | 306 | ||
AK088047.1 | - | 3 | 277 | |||
AK150222.1 | - | 3 | 267 | |||
AK161309.1 | - | 3 | 295 | |||
BC006841.1 | - | 3 | 296 | 3598157 | ||
CF163926.1 | - | 3 | 336 |