Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028662.2 | NP_082938.1 | 3 | 1029 | ||
GenBank mRNA | AF414190.1 | - | 4 | 702 | ||
AK090329.1 | - | 3 | 1218 | |||
AK144403.1 | - | 4 | 725 | |||
AK152362.1 | - | 4 | 703 | |||
AK154868.1 | - | 4 | 688 | |||
AK155551.1 | - | 4 | 735 | |||
AK167343.1 | - | 3 | 1030 | |||
AK186794.1 | - | 1 | 60 | |||
BC025875.1 | - | 4 | 701 | 5137689 | ||
BC036992.1 | - | 4 | 709 | 5003855 |