Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300899.1 | NP_001287828.1 | 12 | 1446 | ||
NM_004999.3 | NP_004990.3 | 12 | 1430 | |||
XM_005248719.3 | XP_005248776.1 | 12 | 1446 | |||
XM_005248720.3 | XP_005248777.1 | 12 | 1446 | |||
XM_005248721.3 | XP_005248778.1 | 12 | 1446 | |||
XM_005248722.3 | XP_005248779.1 | 12 | 1446 | |||
XM_005248724.3 | XP_005248781.1 | 12 | 1446 | |||
XM_005248726.3 | XP_005248783.1 | 12 | 1446 | |||
XM_017010899.1 | XP_016866388.1 | 12 | 1446 | |||
GenBank mRNA | AB002387.1 | - | 12 | 1290 | ||
BC146764.1 | - | 12 | 1290 | 9007206 | ||
BX647397.1 | - | 12 | 1421 | |||
U90236.2 | - | 11 | 1271 |