Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028710.3 | NP_082986.3 | 4 | 1053 | ||
GenBank mRNA | AK018132.2 | - | 1 | 470 | ||
AK158726.1 | - | 4 | 1054 | |||
AK173082.1 | - | 1 | 152 | |||
BC084731.1 | - | 1 | 743 | 6813261 |