Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028718.2 | NP_082994.1 | 2, 3 | 298 | ||
GenBank mRNA | AY613437.1 | - | 2, 3 | 301 | ||
BC046538.1 | - | 2, 3 | 301 | |||
BC117868.2 | - | 2, 3 | 272 | 40095476 | ||
BY739359.1 | - | 2, 3 | 298 |