Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028735.3 | NP_083011.2 | 24 | 3302 | ||
GenBank mRNA | AK015017.1 | - | 24 | 3262 | ||
AK029952.1 | - | 24 | 3306 | |||
AK133304.1 | - | 24 | 3274 | |||
BC133699.1 | - | 24 | 3260 | 40098338 | ||
DQ011665.1 | - | 24 | 3306 |