Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277292.1 | NP_001264221.1 | 22, 23 | 3287 | ||
NM_001277293.1 | NP_001264222.1 | 23, 24 | 3443 | |||
NM_001277294.1 | NP_001264223.1 | 22, 23 | 3144 | |||
NM_001277295.1 | NP_001264224.1 | 12, 13 | 1884 | |||
NM_028736.2 | NP_083012.1 | 24, 25 | 3345 | |||
NM_130891.2 | NP_570961.1 | 21, 22 | 3063 | |||
NM_133442.2 | NP_597699.1 | 21, 22 | 3206 | |||
GenBank mRNA | AB051560.1 | - | 21, 22 | 2940 | ||
AB051561.1 | - | 24, 25 | 3222 | |||
AB051562.1 | - | 21, 22 | 2926 | |||
AK016420.1 | - | 12, 13 | 1882 | |||
AK029845.1 | - | 21, 22 | 3605 | |||
AK029905.1 | - | 12, 13 | 1885 | |||
AK043821.1 | - | 1, 2 | 27 | |||
AK220553.1 | - | 22, 23 | 3001 | |||
AY255674.1 | - | 12, 13 | 1700 | |||
AY294283.1 | - | 12, 13 | 1681 | |||
BC067398.1 | - | 22, 23 | 3144 | 30096759 | ||
BC072632.1 | - | 23, 24 | 3443 | 6843802 |