Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164362.1 | NP_001157834.1 | 4 | 721 | ||
NM_028293.1 | NP_082569.1 | 3 | 345 | |||
NM_028760.2 | NP_083036.2 | 4 | 697 | |||
GenBank mRNA | AK004655.1 | - | 4 | 723 | ||
AK012311.1 | - | 4 | 494 | |||
AK028216.1 | - | 3 | 331 | |||
AK088548.1 | - | 4 | 404 | |||
BC026966.1 | - | 1 | 53 | |||
BC031396.1 | - | 4 | 650 | 4950530 | ||
CJ074252.1 | - | 3 | 348 |