Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256096.1 | NP_001243025.1 | 4 | 799 | ||
NM_001256097.1 | NP_001243026.1 | 5 | 877 | |||
NM_001256098.1 | NP_001243027.1 | 4 | 799 | |||
NM_023742.2 | NP_076231.1 | 4 | 799 | |||
GenBank mRNA | AB015423.1 | - | 4 | 722 | ||
AB015424.1 | - | 4 | 722 | |||
AK012137.1 | - | 4 | 795 | |||
AK044105.1 | - | 5 | 834 | |||
AK054443.1 | - | 4 | 789 | |||
BC024925.1 | - | 4 | 741 | 3711019 |