Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028862.3 | NP_083138.2 | 9 | 1598 | ||
GenBank mRNA | AK014408.1 | - | 9 | 1599 | ||
AK043002.1 | - | 9 | 1585 | |||
BC040799.1 | - | 8 | 1274 | 3988157 | ||
BC051064.1 | - | 2 | 301 | |||
BC055485.1 | - | 3 | 382 |