Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028863.1 | NP_083139.1 | 2 | 152 | ||
GenBank mRNA | AK002936.1 | - | 2 | 151 | ||
AK019988.1 | - | 1 | 319 | |||
BC022589.1 | - | 2 | 136 | 4037176 | ||
BY125913.1 | - | 2 | 145 |