Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 2, 3 | 587 | ||
NM_022892.1 | NP_075043.1 | 2, 3 | 587 | |||
GenBank mRNA | AB048534.1 | - | 2, 3 | 587 | ||
U19251.1 | - | 2, 3 | 160 |