Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023750.2 | NP_076239.1 | 5 | 485 | ||
GenBank mRNA | AK014610.1 | - | 5 | 441 | ||
AK034259.1 | - | 5 | 380 | |||
AK083748.1 | - | 5 | 425 | |||
AK145426.1 | - | 5 | 486 | |||
AK158422.1 | - | 5 | 435 | |||
BC008540.1 | - | 5 | 433 | 3495627 | ||
U29503.1 | - | 3 | 195 |