Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026665.4 | NP_080941.3 | 7, 8 | 1025 | ||
GenBank mRNA | AK014862.1 | - | 4, 5 | 658 | ||
AK014873.1 | - | 7, 8 | 972 | |||
AK014982.1 | - | 5, 6 | 483 | |||
AK016484.1 | - | 7, 8 | 809 | |||
AK019414.1 | - | 7, 8 | 905 | |||
AK028458.1 | - | 7, 8 | 853 | |||
AK167523.1 | - | 7, 8 | 910 | |||
AY225093.1 | - | 7, 8 | 818 | |||
AY251192.1 | - | 7, 8 | 970 | |||
BC025588.1 | - | 6, 7 | 805 | |||
BC050785.1 | - | 5, 6 | 536 | 6703205 | ||
BC056968.1 | - | 7, 8 | 1269 |