Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 8 | 939 | ||
AK016530.1 | - | 8 | 1086 | |||
AK144082.1 | - | 8 | 1734 | |||
AK161011.1 | - | 8 | 1845 | |||
AK172276.1 | - | 8 | 1732 | |||
BC060140.1 | - | 8 | 1051 | |||
BC118971.1 | - | 8 | 934 |