Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004538.5 | NP_004529.2 | 1 | 1793 | ||
GenBank mRNA | AK090772.1 | - | 2 | 1599 | ||
AK095682.1 | - | 2 | 1650 | |||
AK223621.1 | - | 1 | 1724 | |||
AK315176.1 | - | 1 | 1686 | |||
BC034954.2 | - | 1 | 1681 | 4823724 | ||
D50370.1 | - | 1 | 1679 |