Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 27 | 3195 | ||
NM_001165984.1 | NP_001159456.1 | 27 | 3155 | |||
NM_028475.2 | NP_082751.1 | 27 | 3180 | |||
NM_153489.2 | NP_705693.2 | 26 | 3117 | |||
GenBank mRNA | AK016538.1 | - | 20 | 2447 | ||
AK036526.1 | - | 9 | 900 | |||
AK048286.1 | - | 27 | 3196 | |||
AK050956.1 | - | 3 | 122 | |||
AK162166.1 | - | 12 | 1208 | |||
BC023906.1 | - | 26 | 3108 | 5321969 | ||
BC029075.1 | - | 27 | 3170 | 4527901 | ||
BC050910.1 | - | 26 | 3102 | 6417527 |