Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 18 | 2193 | ||
NM_001165984.1 | NP_001159456.1 | 18 | 2153 | |||
NM_001165985.1 | NP_001159457.1 | 18 | 2211 | |||
NM_001165986.1 | NP_001159458.1 | 18 | 2186 | |||
NM_001165987.1 | NP_001159459.1 | 17 | 2118 | |||
NM_001165988.1 | NP_001159460.1 | 17 | 2093 | |||
NM_028475.2 | NP_082751.1 | 18 | 2178 | |||
NM_153489.2 | NP_705693.2 | 17 | 2118 | |||
GenBank mRNA | AF276967.1 | - | 17 | 2053 | ||
AK016538.1 | - | 11 | 1445 | |||
AK030218.1 | - | 18 | 2119 | |||
AK031934.1 | - | 17 | 2098 | |||
AK048286.1 | - | 18 | 2194 | |||
AK079890.1 | - | 1 | 423 | |||
AK079895.1 | - | 18 | 2150 | |||
AK086314.1 | - | 3 | 1579 | |||
AK088702.1 | - | 18 | 2209 | |||
AK122210.1 | - | 5 | 556 | |||
AK162166.1 | - | 3 | 206 | |||
AK213629.1 | - | 3 | 192 | |||
BC023906.1 | - | 17 | 2109 | 5321969 | ||
BC029075.1 | - | 18 | 2168 | 4527901 | ||
BC050910.1 | - | 17 | 2100 | 6417527 |