Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144535.4 | NP_653118.3 | 2 | 612 | ||
GenBank mRNA | AK016540.1 | - | 2 | 559 | ||
AK028927.1 | - | 2 | 565 | |||
AK030928.1 | - | 2 | 539 | |||
AK031527.1 | - | 2 | 1011 | |||
AK036454.1 | - | 2 | 537 | |||
AK036465.1 | - | 2 | 544 | |||
AK042544.1 | - | 2 | 534 | |||
AK051372.1 | - | 2 | 559 | |||
AK164148.1 | - | 2 | 383 | |||
AK170232.1 | - | 2 | 389 | |||
AK177699.1 | - | 1 | 103 | |||
AK211895.1 | - | 2 | 289 | |||
BC023094.1 | - | 2 | 612 | 4017596 |