Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028959.3 | NP_083235.3 | 5 | 611 | ||
GenBank mRNA | AK017134.1 | - | 5 | 603 | ||
AK034892.1 | - | 5 | 604 | |||
BC060686.1 | - | 5 | 609 | |||
BC080681.1 | - | 5 | 567 | |||
BC100549.1 | - | 4 | 461 |