Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028976.2 | NP_083252.1 | 9 | 1546 | ||
GenBank mRNA | AK154986.1 | - | 9 | 1523 | ||
AK170627.1 | - | 9 | 1510 | |||
AK186298.1 | - | 1 | 256 | |||
AK197927.1 | - | 1 | 170 | |||
AK200534.1 | - | 1 | 171 | |||
BC012251.1 | - | 9 | 1546 | 4166619 |