Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167939.1 | NP_001161411.1 | 5 | 606 | ||
NM_028993.4 | NP_083269.4 | 5 | 606 | |||
GenBank mRNA | AK018662.1 | - | 5 | 574 | ||
AK034837.1 | - | 5 | 568 | |||
AK039196.1 | - | 5 | 572 | |||
AK049277.1 | - | 5 | 567 | |||
AK081967.1 | - | 5 | 607 | |||
AK162318.1 | - | 5 | 573 | |||
AK164005.1 | - | 5 | 573 | |||
BC125568.1 | - | 5 | 606 | 40130083 | ||
BC132139.1 | - | 5 | 606 | 40130416 | ||
BC144990.1 | - | 5 | 606 | 9053538 |