Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000433.3 | NP_000424.2 | 5 | 821 | ||
NM_001127651.2 | NP_001121123.1 | 6 | 658 | |||
NM_001190794.1 | NP_001177723.1 | 4 | 686 | |||
XM_011509580.1 | XP_011507882.1 | 6 | 659 | |||
XM_011509581.1 | XP_011507883.1 | 6 | 646 | |||
GenBank mRNA | AB209647.1 | - | 5 | 701 | ||
BP193903.1 | - | 6 | 505 | |||
KU178216.1 | - | 5 | 546 | |||
M32011.1 | - | 6 | 613 |