Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081160.1 | NP_001074629.1 | 15 | 3109 | ||
GenBank mRNA | AK004706.1 | - | 8 | 1119 | ||
DQ788983.1 | - | 3538 |