Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029091.2 | NP_083367.1 | 15 | 1854 | ||
GenBank mRNA | AK004774.1 | - | 15 | 1855 | ||
AK170793.1 | - | 15 | 1838 | |||
AK219585.1 | - | 1 | 26 |