Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205157.1 | NP_001192086.1 | 3, 4 | 325 | ||
NM_001205158.1 | NP_001192087.1 | 3, 4 | 315 | |||
NM_138303.2 | NP_612176.1 | 2, 3 | 463 | |||
GenBank mRNA | AA967152.1 | - | 2, 3 | 262 | ||
AK004959.1 | - | 2, 3 | 249 | |||
AK033705.1 | - | 3, 4 | 274 | |||
AK133631.1 | - | 3, 4 | 285 | |||
BC002282.1 | - | 2, 3 | 378 | 3497525 | ||
BC003289.1 | - | 3, 4 | 256 | 3583087 |