Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027554.2 | NP_081830.2 | 9 | 2794 | ||
GenBank mRNA | AK014740.1 | - | 3 | 898 | ||
AK028904.1 | - | 9 | 2773 | |||
AK052219.1 | - | 9 | 2817 | |||
AK054116.1 | - | 9 | 2796 | |||
AK129464.1 | - | 9 | 2750 | |||
AK142200.1 | - | 9 | 2789 | |||
BC054404.1 | - | 9 | 2794 | 6494448 | ||
BC057122.1 | - | 9 | 2784 | 6834543 | ||
BC058784.1 | - | 9 | 2722 | 6310341 | ||
BC066786.1 | - | 9 | 2738 |