Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029106.2 | NP_083382.2 | 1 | 380 |
Gene Symbol: Gm21637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270685.1 | NP_001257614.1 | 1 | 380 | ||
GenBank mRNA | BC117726.1 | - | 1 | 380 | 40089629 |
Gene Symbol: Gm2837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117727.1 | - | 1 | 380 | 40089638 |
Gene Symbol: Spin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243002.1 | NP_001229931.1 | 2 | 589 | ||
GenBank mRNA | AK015110.1 | - | 2 | 590 |