Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191051.1 | NP_001177980.1 | 3 | 760 | ||
NM_175215.4 | NP_780424.1 | 3 | 423 | |||
GenBank mRNA | AK033681.1 | - | 3 | 360 | ||
AK143241.1 | - | 3 | 761 | |||
AK172203.1 | - | 4 | 523 | |||
BC030310.1 | - | 4 | 1230 | |||
BC051181.1 | - | 1 | 208 | |||
BC057459.1 | - | 3 | 370 | 4504057 | ||
BC094328.1 | - | 4 | 773 |