Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173745.5 | NP_776106.1 | 2 | 468 | ||
GenBank mRNA | AK015917.1 | - | 2 | 452 | ||
AK079664.1 | - | 2 | 464 | |||
AK081916.1 | - | 2 | 427 | |||
BC020036.1 | - | 2 | 462 | 3990751 |