Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133803.2 | NP_598564.2 | 18 | 2129 | ||
GenBank mRNA | AK031020.1 | - | 18 | 2094 | ||
AK087994.1 | - | 18 | 2093 | |||
AK150101.1 | - | 18 | 2089 | |||
AK150126.1 | - | 18 | 2083 | |||
AK166842.1 | - | 18 | 2094 | |||
AK168195.1 | - | 18 | 2082 | |||
AK185970.1 | - | 2 | 190 | |||
AK200466.1 | - | 2 | 158 | |||
AK207406.1 | - | 2 | 52 | |||
BC004600.1 | - | 17 | 2071 | 3584924 | ||
BC066817.1 | - | 18 | 2080 | 30429618 |