Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133803.2 | NP_598564.2 | 10, 11 | 1266 | ||
GenBank mRNA | AK031020.1 | - | 10, 11 | 1231 | ||
AK087994.1 | - | 10, 11 | 1230 | |||
AK149853.1 | - | 10, 11 | 1231 | |||
AK150101.1 | - | 10, 11 | 1226 | |||
AK150126.1 | - | 10, 11 | 1220 | |||
AK166842.1 | - | 10, 11 | 1231 | |||
AK168195.1 | - | 10, 11 | 1219 | |||
BC004600.1 | - | 9, 10 | 1208 | 3584924 | ||
BC066817.1 | - | 10, 11 | 1217 | 30429618 |