Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pelp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029231.4 | NP_083507.3 | 16 | 2664 | ||
GenBank mRNA | AK005027.1 | - | 16 | 2662 | ||
AK128966.1 | - | 16 | 2664 | |||
BC016444.1 | - | 6 | 1481 | 4459875 | ||
BC057987.1 | - | 10 | 1892 | |||
BC090620.1 | - | 16 | 2658 | 30699968 |