Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcdin3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029236.2 | NP_083512.2 | 1 | 135 | ||
GenBank mRNA | AK016154.1 | - | 1 | 137 | ||
BC016225.1 | - | 1 | 114 | 4486953 | ||
BC078455.1 | - | 1 | 113 | |||
BY262678.1 | - | 1 | 136 | |||
CO427236.1 | - | 1 | 95 |