Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005003.2 | NP_004994.1 | 3, 4 | 405 | ||
XM_011545856.2 | XP_011544158.2 | 4, 5 | 509 | |||
GenBank mRNA | AF087660.1 | - | 3, 4 | 401 | ||
AK311877.1 | - | 3, 4 | 400 | |||
BC058920.1 | - | 3, 4 | 395 | 6167387 | ||
BI603093.1 | - | 3, 4 | 444 |