Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008506.2 | NP_001008506.2 | 13 | 1815 | ||
NM_001160107.1 | NP_001153579.1 | 12 | 1740 | |||
NM_001160108.1 | NP_001153580.1 | 5 | 1009 | |||
NM_029334.2 | NP_083610.2 | 15 | 2208 | |||
GenBank mRNA | AK006009.1 | - | 5 | 973 | ||
AK048046.1 | - | 15 | 2174 | |||
AK050226.1 | - | 15 | 2189 | |||
AK076066.1 | - | 11 | 1609 | |||
AK160937.1 | - | 12 | 1726 | |||
BC024824.1 | - | 12 | 1710 | 5355748 | ||
BC024856.1 | - | 8 | 830 | |||
BC026610.1 | - | 9 | 1280 | |||
BC034760.1 | - | 15 | 2141 |