Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029355.2 | NP_083631.1 | 3 | 291 | ||
GenBank mRNA | AF525156.1 | - | 3 | 291 | ||
AK005458.1 | - | 3 | 292 | |||
BC116772.1 | - | 3 | 268 | 40126091 | ||
BC119183.1 | - | 3 | 268 | 8733932 |