Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081241.2 | NP_001074710.2 | 19 | 3471 | ||
GenBank mRNA | AK010063.1 | - | 1 | 56 | ||
AK052316.1 | - | 20 | 3352 | |||
AK122573.1 | - | 19 | 3445 | |||
BC006820.1 | - | 2 | 257 | |||
BC030451.1 | - | 9 | 2534 | 5400266 | ||
BC079880.1 | - | 19 | 3385 | 6852251 |